Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107165.1 | NP_001100635.1 | 1 - 2 | 524 | 122 | |
XM_008772381.1 | XP_008770603.1 | 2 - 3 | 648 | 122 | ||
XM_008772382.1 | XP_008770604.1 | 2 - 3 | 663 | 122 | ||
XM_008772383.1 | XP_008770605.1 | 2 - 3 | 645 | 122 | ||
XM_008772384.1 | XP_008770606.1 | 2 - 3 | 622 | 122 | ||
XM_008772385.1 | XP_008770607.1 | 3 - 4 | 584 | 122 | ||
XM_008772386.1 | XP_008770608.1 | 3 - 4 | 459 | 122 | ||
XM_008772387.1 | XP_008770609.1 | 2 - 3 | 477 | 122 | ||
XM_008772388.1 | XP_008770610.1 | 2 - 3 | 594 | 122 | ||
XM_008772389.1 | XP_008770611.1 | 2 - 3 | 571 | 122 | ||
XM_008772390.1 | XP_008770612.1 | 2 - 3 | 590 | 122 | ||
XM_008772391.1 | XP_008770613.1 | 2 - 3 | 575 | 122 | ||
XM_008772392.1 | XP_008770614.1 | 2 - 3 | 289 | 122 | ||
XM_008772393.1 | XP_008770615.1 | 2 - 3 | 598 | 122 | ||
XM_008772394.1 | XP_008770616.1 | 2 - 3 | 378 | 122 | ||
XM_008772395.1 | XP_008770617.1 | 2 - 3 | 371 | 122 | ||
XM_008772396.1 | XP_008770618.1 | 2 - 3 | 451 | 122 | ||
XM_008772397.1 | XP_008770619.1 | 2 - 3 | 404 | 122 | ||
XM_008772398.1 | XP_008770620.1 | 2 - 3 | 352 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |