Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106670.1 | NP_001100140.1 | 7 - 8 | 1134 | 85 | |
GenBank mRNA | FQ211202.1 | - | 7 - 8 | 1213 | 85 | |
FQ211244.1 | - | 7 - 8 | 1213 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |