Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015015.1 | NP_001015015.1 | 3 - 4 | 738 | 92 | |
GenBank mRNA | BC092574.1 | - | 3 - 4 | 738 | 7113947 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |