Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238423.2 | XP_006238485.1 | 26 - 27 | 3733 | 83 | |
XM_006238424.2 | XP_006238486.1 | 26 - 27 | 3739 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |