Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147146.2 | NP_671487.1 | 2 - 3 | 265 | 186 | |
XM_008772869.1 | XP_008771091.1 | 2 - 3 | 397 | 186 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |