Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007601.2 | NP_001007602.1 | 1 - 2 | 41 | 62 | |
XM_006256414.1 | XP_006256476.1 | 2 - 3 | 299 | 62 | ||
GenBank mRNA | FM060466.1 | - | 1 - 2 | 41 | 62 | |
M83740.1 | - | 1 - 2 | 237 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |