Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024743.1 | NP_001019914.1 | 4 - 5 | 695 | 114 | |
XM_006251538.1 | XP_006251600.1 | 4 - 5 | 647 | 114 | ||
GenBank mRNA | BC097369.1 | - | 4 - 5 | 695 | 7443949 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |