Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108367.1 | NP_001101837.1 | 5 - 6 | 1342 | 110 | |
GenBank mRNA | FQ211609.1 | - | 2 - 3 | 620 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |