Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108367.1 | NP_001101837.1 | 6 - 7 | 1458 | 106 | |
GenBank mRNA | FQ211609.1 | - | 3 - 4 | 736 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |