Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004214.1 | NP_001004214.1 | 5 - 6 | 516 | 70 | |
XM_006253867.2 | XP_006253929.1 | 5 - 6 | 767 | 70 | ||
XM_006253868.2 | XP_006253930.1 | 4 - 5 | 525 | 70 | ||
GenBank mRNA | BC079157.1 | - | 5 - 6 | 516 | 7128990 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |