Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271330.1 | NP_001258259.1 | 4 - 5 | 651 | 86 | |
GenBank mRNA | FM067602.1 | - | 4 - 5 | 417 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |