Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008556.1 | NP_001008556.1 | 8 - 9 | 637 | 67 | |
XM_008771534.1 | XP_008769756.1 | 7 - 8 | 1352 | 67 | ||
XM_008771535.1 | XP_008769757.1 | 8 - 9 | 736 | 67 | ||
GenBank mRNA | AY823319.1 | - | 8 - 9 | 637 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |