Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170429.1 | NP_001163900.1 | 1 - 2 | 303 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |