Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105743.1 | NP_001099213.1 | 1 - 2 | 472 | 138 | |
GenBank mRNA | AB001453.1 | - | 1 - 2 | 686 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |