Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013136.1 | NP_037268.1 | 3 - 4 | 513 | 183 | |
GenBank mRNA | BC078887.1 | - | 3 - 4 | 501 | 7111717 | 183 |
M35862.1 | - | 3 - 4 | 513 | 183 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |