Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107345.1 | NP_001100815.1 | 2 - 3 | 749 | 71 | |
XM_008771570.1 | XP_008769792.1 | 2 - 3 | 486 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |