Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199085.2 | NP_954516.1 | 2 - 3 | 267 | 64 | |
XM_006253869.1 | XP_006253931.1 | 2 - 3 | 265 | 64 | ||
XM_006253870.2 | XP_006253932.1 | 2 - 3 | 560 | 64 | ||
XM_006253871.2 | XP_006253933.1 | 2 - 3 | 380 | 64 | ||
XM_008771596.1 | XP_008769818.1 | 2 - 3 | 478 | 64 | ||
GenBank mRNA | BC060594.1 | - | 2 - 3 | 271 | 6887824 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |