Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107348.1 | NP_001100818.1 | 11 - 12 | 4334 | 60 | |
XM_006253816.2 | XP_006253878.1 | 10 - 11 | 4312 | 60 | ||
XM_008771571.1 | XP_008769793.1 | 9 - 10 | 4066 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |