Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107348.1 | NP_001100818.1 | 4 - 5 | 480 | 98 | |
XM_006253812.2 | XP_006253874.1 | 3 - 4 | 458 | 98 | ||
XM_006253813.2 | XP_006253875.1 | 2 - 3 | 212 | 98 | ||
XM_006253814.2 | XP_006253876.1 | 3 - 4 | 827 | 98 | ||
XM_006253815.2 | XP_006253877.1 | 3 - 4 | 1424 | 98 | ||
XM_006253816.2 | XP_006253878.1 | 3 - 4 | 458 | 98 | ||
XM_008771571.1 | XP_008769793.1 | 2 - 3 | 212 | 98 | ||
GenBank mRNA | FQ222841.1 | - | 3 - 4 | 273 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |