Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012052.1 | NP_001012052.1 | 3 - 4 | 466 | 125 | |
XM_006253808.2 | XP_006253870.1 | 3 - 4 | 453 | 125 | ||
XM_006253809.2 | XP_006253871.1 | 2 - 3 | 144 | 125 | ||
XM_006253810.2 | XP_006253872.1 | 3 - 4 | 483 | 125 | ||
XM_006253811.2 | XP_006253873.1 | 3 - 4 | 567 | 125 | ||
GenBank mRNA | BC081934.1 | - | 3 - 4 | 466 | 7114574 | 125 |
FQ211932.1 | - | 3 - 4 | 572 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |