Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012052.1 | NP_001012052.1 | 5 - 6 | 2053 | 140 | |
XM_006253808.2 | XP_006253870.1 | 5 - 6 | 2040 | 140 | ||
XM_006253809.2 | XP_006253871.1 | 4 - 5 | 1731 | 140 | ||
GenBank mRNA | BC081934.1 | - | 5 - 6 | 2053 | 7114574 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |