Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108406.1 | NP_001101876.1 | 10 - 11 | 997 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |