Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129879.2 | NP_001123351.1 | 2 - 3 | 348 | 110 | |
GenBank mRNA | BC167096.1 | - | 2 - 3 | 275 | 8362489 | 110 |
BE113476.1 | - | 1 - 2 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |