Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011922.1 | NP_001011922.1 | 4 - 5 | 759 | 103 | |
XM_006253786.2 | XP_006253848.1 | 4 - 5 | 922 | 103 | ||
XM_008771569.1 | XP_008769791.1 | 4 - 5 | 1575 | 103 | ||
GenBank mRNA | BC085812.1 | - | 4 - 5 | 759 | 7121465 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |