Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01435436_m1
See other RABEPK GE Assays ›
Gene Symbol:
Rabepk
Gene Name:
Rab9 effector protein with kelch motifs
Gene Aliases:
9530020d24rik, RGD1310612
Chromosome Location:
Chr.3: 13845352 - 13865843 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01435436_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Rabepk
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001024871.1
NP_001020042.1
5 - 6
797
126
XM_006233955.1
XP_006234017.1
4 - 5
844
126
XM_006233957.2
XP_006234019.1
4 - 5
452
126
XM_006233958.1
XP_006234020.1
4 - 5
711
126
XM_006233963.1
XP_006234025.1
4 - 5
760
126
XM_006233964.2
XP_006234026.1
4 - 5
369
126
XM_006233966.1
XP_006234028.1
4 - 5
756
126
XM_006233967.1
XP_006234029.1
4 - 5
623
126
XM_008761724.1
XP_008759946.1
6 - 7
994
126
XM_008761726.1
XP_008759948.1
5 - 6
525
126
XM_008761727.1
XP_008759949.1
4 - 5
871
126
XM_008761728.1
XP_008759950.1
4 - 5
480
126
XM_008761729.1
XP_008759951.1
4 - 5
738
126
XM_008761730.1
XP_008759952.1
5 - 6
796
126
XM_008761731.1
XP_008759953.1
5 - 6
441
126
XM_008761733.1
XP_008759955.1
3 - 4
762
126
XM_008761736.1
XP_008759958.1
3 - 4
414
126
XM_008761738.1
XP_008759960.1
4 - 5
627
126
XM_008761739.1
XP_008759961.1
3 - 4
673
126
GenBank mRNA
BC097415.1
-
5 - 6
797
7454340
126
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300002917.rn.1
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Rattus norvegicus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Rabepk
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0300022641.rn.1
TC0300002917.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0300022639.rn.1
TC0300002917.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0