Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145677.1 | NP_663710.1 | 2 - 3 | 387 | 67 | |
XM_006233931.2 | XP_006233993.1 | 2 - 3 | 508 | 67 | ||
XM_006233932.2 | XP_006233994.1 | 2 - 3 | 508 | 67 | ||
XM_006233933.2 | XP_006233995.1 | 2 - 3 | 1160 | 67 | ||
XM_006233934.2 | XP_006233996.1 | 2 - 3 | 1160 | 67 | ||
XM_006233935.2 | XP_006233997.1 | 2 - 3 | 1724 | 67 | ||
XM_006233937.2 | XP_006233999.1 | 2 - 3 | 744 | 67 | ||
GenBank mRNA | AY043169.1 | - | 2 - 3 | 387 | 67 | |
FQ211394.1 | - | 2 - 3 | 403 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |