Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsb66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233762.1 | XP_006233824.1 | 1 - 2 | 213 | 114 | |
XM_006233766.1 | XP_006233828.1 | 1 - 2 | 213 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |