Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108570.1 | NP_001102040.1 | 1 - 2 | 59 | 115 | |
XM_006233614.1 | XP_006233676.1 | 2 - 3 | 198 | 115 | ||
XM_006233615.1 | XP_006233677.1 | 3 - 4 | 1518 | 115 | ||
XM_006233616.2 | XP_006233678.1 | 3 - 4 | 401 | 115 | ||
XM_006233617.2 | XP_006233679.1 | 3 - 4 | 614 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |