Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031044.3 | NP_112306.1 | 4 - 5 | 440 | 92 | |
GenBank mRNA | BC087635.1 | - | 4 - 5 | 631 | 7319794 | 92 |
D10693.1 | - | 4 - 5 | 440 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |