Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033884.1 | NP_001029056.1 | 3 - 4 | 750 | 112 | |
GenBank mRNA | BC161820.1 | - | 3 - 4 | 715 | 9088929 | 112 |
CB611468.1 | - | 2 - 3 | 235 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |