Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107814.1 | NP_001101284.1 | 4 - 5 | 438 | 122 | |
XM_008761586.1 | XP_008759808.1 | 2 - 3 | 1089 | 122 | ||
XM_008761587.1 | XP_008759809.1 | 3 - 4 | 732 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |