Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079941.1 | NP_001073410.1 | 7 - 8 | 1318 | 60 | |
XM_006233835.2 | XP_006233897.1 | 7 - 8 | 1327 | 60 | ||
GenBank mRNA | BC129109.1 | - | 7 - 8 | 1318 | 7105121 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |