Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171802.1 | NP_001165273.1 | 3 - 4 | 356 | 94 | |
XM_006237906.2 | XP_006237968.1 | 3 - 4 | 367 | 94 | ||
GenBank mRNA | BC089993.1 | - | 3 - 4 | 326 | 7319652 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |