Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134467.1 | NP_604462.1 | 6 - 7 | 889 | 149 | |
XM_006238012.2 | XP_006238074.1 | 6 - 7 | 1139 | 149 | ||
XM_006238014.2 | XP_006238076.1 | 6 - 7 | 1037 | 149 | ||
XM_006238015.2 | XP_006238077.1 | 5 - 6 | 989 | 149 | ||
XM_006238016.2 | XP_006238078.1 | 5 - 6 | 1236 | 149 | ||
XM_006238017.2 | XP_006238079.1 | 5 - 6 | 2009 | 149 | ||
XM_006238018.2 | XP_006238080.1 | 6 - 7 | 1077 | 149 | ||
XM_006238020.2 | XP_006238082.1 | 6 - 7 | 882 | 149 | ||
XM_008763664.1 | XP_008761886.1 | 5 - 6 | 1663 | 149 | ||
XM_008763665.1 | XP_008761887.1 | 5 - 6 | 1576 | 149 | ||
XM_008763666.1 | XP_008761888.1 | 5 - 6 | 1561 | 149 | ||
XM_008763667.1 | XP_008761889.1 | 7 - 8 | 1032 | 149 | ||
XM_008763668.1 | XP_008761890.1 | 6 - 7 | 975 | 149 | ||
GenBank mRNA | AF480444.1 | - | 6 - 7 | 889 | 149 | |
BC083548.1 | - | 5 - 6 | 804 | 7134112 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |