Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024900.1 | NP_001020071.1 | 1 - 2 | 173 | 100 | |
GenBank mRNA | BC097492.1 | - | 1 - 2 | 173 | 7461463 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |