Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749927.3 | XP_003749975.1 | 3 - 4 | 2950 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |