Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108657.1 | NP_001102127.1 | 4 - 5 | 836 | 62 | |
XM_006237901.2 | XP_006237963.1 | 5 - 6 | 969 | 62 | ||
XM_006237903.2 | XP_006237965.1 | 4 - 5 | 741 | 62 | ||
XM_006237904.2 | XP_006237966.1 | 4 - 5 | 799 | 62 | ||
XM_008763548.1 | XP_008761770.1 | 2 - 3 | 2247 | 62 | ||
XM_008763549.1 | XP_008761771.1 | 4 - 5 | 6132 | 62 | ||
XM_008763550.1 | XP_008761772.1 | 5 - 6 | 665 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |