Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012101.2 | NP_001012101.2 | 5 - 6 | 795 | 89 | |
XM_006238058.2 | XP_006238120.1 | 4 - 5 | 2350 | 89 | ||
GenBank mRNA | BC079209.1 | - | 5 - 6 | 748 | 7131801 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |