Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749909.3 | XP_003749957.1 | 2 - 3 | 1022 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |