Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343571.7 | XP_343572.4 | 6 - 7 | 1584 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |