Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106914.1 | NP_001100384.1 | 7 - 8 | 4235 | 80 | |
XM_006244965.2 | XP_006245027.1 | 5 - 6 | 2284 | 80 | ||
XM_006244966.2 | XP_006245028.1 | 5 - 6 | 836 | 80 | ||
XM_006244967.2 | XP_006245029.1 | 5 - 6 | 1348 | 80 | ||
XM_008767084.1 | XP_008765306.1 | 4 - 5 | 3030 | 80 | ||
XM_008767085.1 | XP_008765307.1 | 4 - 5 | 3148 | 80 | ||
XM_008767086.1 | XP_008765308.1 | 5 - 6 | 841 | 80 | ||
XM_008767087.1 | XP_008765309.1 | 5 - 6 | 770 | 80 | ||
GenBank mRNA | FQ209862.1 | - | 6 - 7 | 1381 | 80 | |
FQ228496.1 | - | 6 - 7 | 1404 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |