Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc40a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133315.2 | NP_579849.2 | 1 - 2 | 332 | 66 | |
GenBank mRNA | AF394785.3 | - | 1 - 2 | 277 | 66 | |
CB808602.1 | - | 1 - 2 | 343 | 66 | ||
U76714.1 | - | 1 - 2 | 347 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |