Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244869.2 | XP_006244931.1 | 11 - 12 | 2138 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |