Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039028.2 | NP_001034117.2 | 6 - 7 | 759 | 76 | |
GenBank mRNA | BC099152.1 | - | 1 - 2 | 353 | 76 | |
BC166803.1 | - | 6 - 7 | 726 | 7938869 | 76 | |
DY315273.1 | - | 3 - 4 | 372 | 76 | ||
FQ228979.1 | - | 6 - 7 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |