Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106901.2 | NP_001100371.2 | 4 - 5 | 1735 | 72 | |
XM_006244756.2 | XP_006244818.1 | 4 - 5 | 1742 | 72 | ||
XM_006244757.2 | XP_006244819.1 | 4 - 5 | 1745 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |