Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133392.1 | NP_596883.1 | 2 - 3 | 288 | 120 | |
XM_006244908.2 | XP_006244970.1 | 2 - 3 | 1057 | 120 | ||
GenBank mRNA | AB070349.1 | - | 2 - 3 | 288 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |