Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109306.1 | NP_001102776.1 | 2 - 3 | 444 | 102 | |
XM_006244931.2 | XP_006244993.1 | 2 - 3 | 1116 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |