Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108216.1 | NP_001101686.1 | 2 - 3 | 252 | 152 | |
XM_006244825.2 | XP_006244887.1 | 2 - 3 | 217 | 152 | ||
XM_008767035.1 | XP_008765257.1 | 2 - 3 | 2661 | 152 | ||
GenBank mRNA | AY724492.1 | - | 2 - 3 | 252 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |