Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nif3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024763.1 | NP_001019934.1 | 3 - 4 | 835 | 84 | |
XM_006244958.2 | XP_006245020.1 | 5 - 6 | 976 | 84 | ||
XM_006244960.2 | XP_006245022.1 | 5 - 6 | 947 | 84 | ||
XM_006244961.2 | XP_006245023.1 | 5 - 6 | 972 | 84 | ||
XM_006244962.2 | XP_006245024.1 | 4 - 5 | 845 | 84 | ||
XM_006244963.2 | XP_006245025.1 | 5 - 6 | 979 | 84 | ||
XM_008767082.1 | XP_008765304.1 | 5 - 6 | 980 | 84 | ||
GenBank mRNA | BC097437.1 | - | 3 - 4 | 835 | 7455562 | 84 |
BC097992.1 | - | 5 - 6 | 908 | 7446502 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |