Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011962.1 | NP_001011962.1 | 13 - 14 | 1507 | 75 | |
XM_006233768.2 | XP_006233830.1 | 10 - 11 | 2175 | 75 | ||
XM_006233769.2 | XP_006233831.1 | 11 - 12 | 1567 | 75 | ||
XM_006233770.2 | XP_006233832.1 | 13 - 14 | 2547 | 75 | ||
XM_006233771.2 | XP_006233833.1 | 9 - 10 | 2041 | 75 | ||
GenBank mRNA | BC085782.1 | - | 13 - 14 | 1507 | 7107449 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |